Assay Details
| Gene Symbol: DAPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348.2 | NP_001339.1 | 5 - 6 | 897 | 84 | |
| XM_005259508.1 | XP_005259565.1 | 5 - 6 | 772 | 84 | ||
| GenBank mRNA | AB007144.1 | - | 4 - 5 | 693 | 84 | |
| AB022341.1 | - | 4 - 5 | 645 | 84 | ||
| AB265224.1 | - | 2 - 3 | 110 | 84 | ||
| AK027590.1 | - | 5 - 6 | 814 | 84 | ||
| AK074799.1 | - | 4 - 5 | 669 | 84 | ||
| AK097643.1 | - | 5 - 6 | 708 | 84 | ||
| BC126430.1 | - | 4 - 5 | 636 | 8992146 | 84 | |
| BC126432.1 | - | 4 - 5 | 634 | 8992148 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cytokinesis
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
apoptotic process
regulation of smooth muscle contraction
regulation of mitotic nuclear division
regulation of mitotic cell cycle
regulation of cell shape
regulation of autophagy
covalent chromatin modification
negative regulation of translation
neuron differentiation
positive regulation of cell migration
intracellular signal transduction
regulation of apoptotic process
positive regulation of apoptotic process
regulation of myosin II filament organization
protein autophosphorylation
regulation of focal adhesion assembly
cellular response to interferon-gamma
positive regulation of canonical Wnt signaling pathway
apoptotic signaling pathway
regulation of cell motility
regulation of actin cytoskeleton reorganization
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein serine/threonine kinase activity
protein binding
ATP binding
protein C-terminus binding
cAMP response element binding protein binding
Rho GTPase binding
identical protein binding
protein homodimerization activity
leucine zipper domain binding
cytokinesis
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
apoptotic process
regulation of smooth muscle contraction
regulation of mitotic nuclear division
regulation of mitotic cell cycle
regulation of cell shape
regulation of autophagy
covalent chromatin modification
negative regulation of translation
neuron differentiation
positive regulation of cell migration
intracellular signal transduction
regulation of apoptotic process
positive regulation of apoptotic process
regulation of myosin II filament organization
protein autophosphorylation
regulation of focal adhesion assembly
cellular response to interferon-gamma
positive regulation of canonical Wnt signaling pathway
apoptotic signaling pathway
regulation of cell motility
regulation of actin cytoskeleton reorganization
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein serine/threonine kinase activity
protein binding
ATP binding
protein C-terminus binding
cAMP response element binding protein binding
Rho GTPase binding
identical protein binding
protein homodimerization activity
leucine zipper domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000385000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000593844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000593844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000593844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000593844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000593844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000594894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000594894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000594894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000596311.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000596311.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000596311.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000596311.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000596311.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000596311.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000596311.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000601824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000601824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000601824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000601824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000601824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | ENST00000601824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009315.hg.1 | NR_030367.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174348.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174348.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174348.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900174348.hg.1 | TC1900009315.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900174348.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174348.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900174348.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000301264.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000385000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000545797.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000593844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000593844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000593844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000593844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000593844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000594894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000594894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000594894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000595279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000596311.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000596311.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000596311.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000596311.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000596311.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000596311.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000596311.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000601824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000601824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000601824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000601824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000601824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | ENST00000601824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NM_001348.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009315.hg.1 | NR_030367.1 | Refseq |

DAPK3
Genomic Map