Assay Details
| Gene Symbol: ADCK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017013174.1 | XP_016868663.1 | 1 - 2 | 138 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017013174.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |

ADCK5
Genomic Map