Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027252.hg.1 |
TC02001686.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02027252.hg.1 |
NR_002201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188207.hg.1 |
ENST00000394290.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188207.hg.1 |
NR_002201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188207.hg.1 |
ENST00000394290.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200188207.hg.1 |
NR_002201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188207.hg.1 |
TC0200012078.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200188208.hg.1 |
NR_002201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188208.hg.1 |
NR_002201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200188208.hg.1 |
TC0200012078.hg.1 |
Clariom D TC |