Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1400155285.hg.1 |
ENST00000335750.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155285.hg.1 |
ENST00000554719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155285.hg.1 |
ENST00000554738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155285.hg.1 |
ENST00000556496.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155285.hg.1 |
NM_004796.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155285.hg.1 |
NR_073547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155285.hg.1 |
ENST00000335750.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155285.hg.1 |
ENST00000554719.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155285.hg.1 |
ENST00000554738.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155285.hg.1 |
ENST00000556496.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155285.hg.1 |
NM_004796.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155285.hg.1 |
NR_073547.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155285.hg.1 |
TC1400007774.hg.1 |
Clariom D TC |