Assay Details
Gene Symbol: LINC01014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046786.1 | 1 - 2 | 176 | 95 | ||
GenBank mRNA | BX105665.1 | - | 1 - 2 | 177 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | ENST00000436432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | ENST00000436432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | ENST00000436432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | ENST00000436432.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | ENST00000441018.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | ENST00000441018.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013905.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300205941.hg.1 | ENST00000436432.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205941.hg.1 | ENST00000441018.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205941.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205941.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205941.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300205941.hg.1 | ENST00000436432.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205941.hg.1 | ENST00000441018.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205941.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205941.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205941.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300205941.hg.1 | TC0300013905.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300205936.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205936.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205936.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300205936.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205936.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205936.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300205936.hg.1 | TC0300013905.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300205938.hg.1 | ENST00000436432.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205938.hg.1 | ENST00000441018.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205938.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205938.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205938.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300205938.hg.1 | ENST00000436432.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205938.hg.1 | ENST00000441018.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205938.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205938.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205938.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300205938.hg.1 | TC0300013905.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300205939.hg.1 | ENST00000436432.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205939.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205939.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205939.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300205939.hg.1 | ENST00000436432.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205939.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205939.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300205939.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300205939.hg.1 | TC0300013905.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | ENST00000436432.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | ENST00000436432.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | ENST00000436432.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | ENST00000436432.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | ENST00000441018.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | ENST00000441018.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | ENST00000455307.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | NR_046786.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013905.hg.1 | NR_046786.1 | Refseq |
