Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017465.hg.1 |
TC0X001291.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017465.hg.1 |
ENST00000317135.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017465.hg.1 |
ENST00000536655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017465.hg.1 |
ENST00000538422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017465.hg.1 |
NM_001243963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017465.hg.1 |
NM_020871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171940.hg.1 |
ENST00000317135.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171940.hg.1 |
ENST00000538422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171940.hg.1 |
NM_001243963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171940.hg.1 |
NM_020871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171940.hg.1 |
ENST00000317135.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171940.hg.1 |
ENST00000538422.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171940.hg.1 |
NM_001243963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171940.hg.1 |
NM_020871.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171940.hg.1 |
TC0X00010559.hg.1 |
Clariom D TC |