Assay Details
Gene Symbol: LRCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243963.1 | NP_001230892.1 | 9 - 10 | 1327 | 127 | |
NM_020871.3 | NP_065922.3 | 9 - 10 | 1327 | 127 | ||
XM_006724724.3 | XP_006724787.2 | 9 - 10 | 1309 | 127 | ||
XM_017029696.1 | XP_016885185.1 | 9 - 10 | 1336 | 127 | ||
XM_017029697.1 | XP_016885186.1 | 9 - 10 | 1337 | 127 | ||
GenBank mRNA | AB040928.1 | - | 5 - 6 | 508 | 127 | |
AK022128.1 | - | 1 - 2 | 83 | 127 | ||
BC125224.1 | - | 9 - 10 | 1320 | 40124466 | 127 | |
BX647193.1 | - | 9 - 10 | 1327 | 127 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X001291.hg
Additional Information:
This assay is in the 5'UTR region of the AK022128.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X017473.hg.1 | TC0X001291.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017473.hg.1 | ENST00000317135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017473.hg.1 | ENST00000538422.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017473.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X017473.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000607417.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171951.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00171951.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00171951.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171951.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171951.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00171951.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00171951.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171951.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00171951.hg.1 | TC0X00010559.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000607417.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
