| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02016975.hg.1 |
TC02001121.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02016975.hg.1 |
ENST00000337386.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02016975.hg.1 |
ENST00000357215.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02016975.hg.1 |
ENST00000409581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02016975.hg.1 |
ENST00000409641.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02016975.hg.1 |
ENST00000545490.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02016975.hg.1 |
NM_005966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200172505.hg.1 |
ENST00000337386.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200172505.hg.1 |
ENST00000409581.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200172505.hg.1 |
ENST00000409641.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200172505.hg.1 |
ENST00000434473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200172505.hg.1 |
NM_005966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200172505.hg.1 |
ENST00000337386.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200172505.hg.1 |
ENST00000409581.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200172505.hg.1 |
ENST00000409641.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200172505.hg.1 |
ENST00000434473.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200172505.hg.1 |
NM_005966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200172505.hg.1 |
TC0200010273.hg.1 |
Clariom D TC |