Assay Details
| Gene Symbol: TRNAU1AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017846.4 | NP_060316.1 | 3 - 4 | 321 | 94 | |
| GenBank mRNA | AK000510.1 | - | 3 - 4 | 253 | 94 | |
| BC000680.2 | - | 3 - 4 | 245 | 3349210 | 94 | |
| BI602122.1 | - | 3 - 4 | 326 | 94 | ||
More Information
Set Membership: |
|

TRNAU1AP
Genomic Map