Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009116.hg.1 |
TC03000474.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009116.hg.1 |
ENST00000394222.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009116.hg.1 |
ENST00000471138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009116.hg.1 |
ENST00000535334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009116.hg.1 |
NM_001174150.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009116.hg.1 |
NM_001174151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03009116.hg.1 |
NM_182896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
ENST00000335438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
ENST00000394222.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
ENST00000471138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
ENST00000475206.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
ENST00000492165.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
ENST00000535334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
NM_001174150.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
NM_001174151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
NM_182896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
ENST00000335438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
ENST00000394222.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
ENST00000471138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
ENST00000475206.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
ENST00000492165.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
ENST00000535334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
NM_001174150.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
NM_001174151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
NM_182896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300157389.hg.1 |
TC0300008046.hg.1 |
Clariom D TC |