Assay Details
| Gene Symbol: HOMER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277077.1 | NP_001264006.1 | 4 - 5 | 1464 | 141 | |
| NM_004272.4 | NP_004263.1 | 7 - 8 | 1854 | 141 | ||
| GenBank mRNA | AF093262.1 | - | 6 - 7 | 792 | 141 | |
| AK223471.1 | - | 7 - 8 | 1116 | 141 | ||
| AK312481.1 | - | 7 - 8 | 1119 | 141 | ||
| AY189939.1 | - | 4 - 5 | 422 | 141 | ||
| AY189940.1 | - | 3 - 4 | 290 | 141 | ||
| BC015502.1 | - | 7 - 8 | 1069 | 3904242 | 141 | |
| BT009846.1 | - | 6 - 7 | 792 | 141 | ||
| Y17829.1 | - | 7 - 8 | 867 | 141 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
phospholipase C-activating G-protein coupled glutamate receptor signaling pathway
chemical synaptic transmission
positive regulation of signal transduction
behavioral response to cocaine
skeletal muscle fiber development
chemical homeostasis within a tissue
response to calcium ion
positive regulation of calcium ion transport
regulation of calcium ion import
regulation of store-operated calcium entry
regulation of cation channel activity
protein binding
G-protein coupled glutamate receptor binding
signaling adaptor activity
ion channel binding
skeletal muscle contraction
phospholipase C-activating G-protein coupled glutamate receptor signaling pathway
chemical synaptic transmission
positive regulation of signal transduction
behavioral response to cocaine
skeletal muscle fiber development
chemical homeostasis within a tissue
response to calcium ion
positive regulation of calcium ion transport
regulation of calcium ion import
regulation of store-operated calcium entry
regulation of cation channel activity
protein binding
G-protein coupled glutamate receptor binding
signaling adaptor activity
ion channel binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178021.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178021.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178021.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178021.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178021.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178021.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178021.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178021.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178021.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178021.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500178021.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178021.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500178021.hg.1 | TC0500011240.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000282260.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000334082.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000460741.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000508576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | ENST00000535690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_001277078.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011240.hg.1 | NM_004272.4 | Refseq |

HOMER1
Genomic Map