Assay Details
| Gene Symbol: PRSS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002773.4 | NP_002764.1 | 4 - 5 | 806 | 89 | |
| GenBank mRNA | AK296656.1 | - | 4 - 5 | 525 | 89 | |
| AK301619.1 | - | 4 - 5 | 609 | 89 | ||
| BC001462.2 | - | 4 - 5 | 717 | 3138532 | 89 | |
| L41351.1 | - | 4 - 5 | 770 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001050.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000564025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000564025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000564025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000564025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000564025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000564025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000564025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000567833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014984.hg.1 | ENST00000317508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014984.hg.1 | ENST00000419768.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014984.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014984.hg.1 | TC16001050.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014985.hg.1 | ENST00000317508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014985.hg.1 | ENST00000419768.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014985.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014985.hg.1 | TC16001050.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014986.hg.1 | ENST00000317508.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014986.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014986.hg.1 | TC16001050.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600177416.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177416.hg.1 | ENST00000567531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177416.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177416.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177416.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177416.hg.1 | ENST00000567531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177416.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177416.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177416.hg.1 | TC1600010006.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000317508.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000564025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000564025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000564025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000564025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000564025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000564025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000564025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000567833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | ENST00000568261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010006.hg.1 | NM_002773.4 | Refseq |

PRSS8
Genomic Map