Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002166.hg.1 |
TC06000241.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002166.hg.1 |
ENST00000244576.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002166.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002167.hg.1 |
TC06000241.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002167.hg.1 |
ENST00000244576.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002167.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002165.hg.1 |
TC06000241.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002165.hg.1 |
ENST00000244576.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002165.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
ENST00000461521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
ENST00000461521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
ENST00000461521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
ENST00000461521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
ENST00000461521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
ENST00000461521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
ENST00000477999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
ENST00000477999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
ENST00000477999.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
ENST00000482767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
ENST00000482767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
ENST00000482767.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
ENST00000461521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
ENST00000461521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
ENST00000461521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
ENST00000461521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
ENST00000461521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
ENST00000461521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
ENST00000477999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
ENST00000477999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
ENST00000477999.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
ENST00000482767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
ENST00000482767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
ENST00000482767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014086.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197783.hg.1 |
ENST00000461521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197783.hg.1 |
ENST00000482767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197783.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197783.hg.1 |
ENST00000461521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197783.hg.1 |
ENST00000482767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197783.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197783.hg.1 |
TC0600014086.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600197781.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197781.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197781.hg.1 |
TC0600014086.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600197782.hg.1 |
ENST00000482767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197782.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197782.hg.1 |
ENST00000482767.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600197782.hg.1 |
NM_001076781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600197782.hg.1 |
TC0600014086.hg.1 |
Clariom D TC |