Assay Details
| Gene Symbol: POLR3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125346.1 | - | 1 | 1412 | 70 | |
| BX537545.1 | - | 1 | 1845 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 320a | MIR320A |
MIRN320, MIRN320A, hsa-mir-320a, mir-320a |
-7.9kb | Anti-sense | Chr. 8 - 22244962 - 22245043 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100507071 | LOC100507071 | 822.0 | Anti-sense | Chr. 8 - 22254576 - 22275162 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

POLR3D
Genomic Map