Assay Details
| Gene Symbol: TAF7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168474.1 | NP_001161946.1 | 8 - 9 | 692 | 64 | |
| NM_024885.3 | NP_079161.3 | 8 - 9 | 860 | 64 | ||
| XM_005262152.1 | XP_005262209.1 | 8 - 9 | 692 | 64 | ||
| XM_006724664.1 | XP_006724727.1 | 8 - 9 | 859 | 64 | ||
| GenBank mRNA | AF285595.1 | - | 8 - 9 | 694 | 64 | |
| AK026810.1 | - | 7 - 8 | 838 | 64 | ||
| AK308261.1 | - | 8 - 9 | 862 | 64 | ||
| BC043391.1 | - | 8 - 9 | 846 | 6154641 | 64 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001215.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
multicellular organism development
spermatogenesis
biological_process
cell differentiation
RNA polymerase II transcriptional preinitiation complex assembly
molecular_function
transcription coactivator activity
transcription factor binding
histone acetyltransferase binding
transcription regulatory region DNA binding
regulation of transcription from RNA polymerase II promoter
multicellular organism development
spermatogenesis
biological_process
cell differentiation
RNA polymerase II transcriptional preinitiation complex assembly
molecular_function
transcription coactivator activity
transcription factor binding
histone acetyltransferase binding
transcription regulatory region DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016353.hg.1 | TC0X001215.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016353.hg.1 | ENST00000324762.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016353.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016353.hg.1 | ENST00000372905.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016353.hg.1 | ENST00000372907.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016353.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016353.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00170336.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00170336.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00170336.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00170336.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00170336.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00170336.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00170336.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00170336.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00170336.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00170336.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00170336.hg.1 | TC0X00010351.hg.1 | Clariom D TC |

TAF7L
Genomic Map