Assay Details
| Gene Symbol: TAF7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK026810.1 | - | 8 - 9 | 952 | 112 | |
More Information
Set Membership: |
|
Panther Classification:
Molecular Function -
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |

TAF7L
Genomic Map