Assay Details
| Gene Symbol: PAPPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002581.4 | NP_002572.2 | 5 - 6 | 2499 | 91 | |
| XM_017014784.1 | XP_016870273.1 | 5 - 6 | 2539 | 91 | ||
| GenBank mRNA | BC078657.1 | - | 5 - 6 | 2129 | 30340336 | 91 |
| X68280.1 | - | 5 - 6 | 1890 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000593.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09006721.hg.1 | TC09000593.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006721.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006721.hg.1 | ENST00000443904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006721.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006722.hg.1 | TC09000593.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006722.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006722.hg.1 | ENST00000443904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006722.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157151.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157151.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157151.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157151.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157151.hg.1 | TC0900008545.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900157150.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157150.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157150.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157150.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157150.hg.1 | TC0900008545.hg.1 | Clariom D TC |

PAPPA
Genomic Map