Assay Details
| Gene Symbol: CCNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031966.3 | NP_114172.1 | 2 - 3 | 445 | 108 | |
| GenBank mRNA | AK289431.1 | - | 2 - 3 | 334 | 108 | |
| AM851053.1 | - | 2 - 3 | 192 | 108 | ||
| BC006510.2 | - | 2 - 3 | 287 | 2963100 | 108 | |
| BI253686.1 | - | 2 - 3 | 445 | 108 | ||
| BT020128.1 | - | 2 - 3 | 192 | 108 | ||
| M25753.1 | - | 2 - 3 | 281 | 108 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000298.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
oocyte maturation
in utero embryonic development
negative regulation of protein phosphorylation
protein complex assembly
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
mitotic spindle organization
mitotic nuclear envelope disassembly
mitotic metaphase plate congression
spermatogenesis
response to mechanical stimulus
negative regulation of gene expression
anaphase-promoting complex-dependent catabolic process
positive regulation of mRNA 3'-end processing
positive regulation of histone phosphorylation
tissue regeneration
response to drug
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of mitotic cell cycle
response to DDT
positive regulation of fibroblast proliferation
digestive tract development
cell division
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
regulation of cell cycle
positive regulation of attachment of spindle microtubules to kinetochore
ventricular cardiac muscle cell development
positive regulation of cardiac muscle cell proliferation
regulation of chromosome condensation
cellular response to iron(III) ion
cellular response to fatty acid
cellular response to organic cyclic compound
cellular response to hypoxia
regulation of mitotic cell cycle spindle assembly checkpoint
histone H3-S10 phosphorylation involved in chromosome condensation
cyclin-dependent protein serine/threonine kinase activity
patched binding
protein binding
protein kinase binding
histone kinase activity
G2/M transition of mitotic cell cycle
oocyte maturation
in utero embryonic development
negative regulation of protein phosphorylation
protein complex assembly
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
mitotic spindle organization
mitotic nuclear envelope disassembly
mitotic metaphase plate congression
spermatogenesis
response to mechanical stimulus
negative regulation of gene expression
anaphase-promoting complex-dependent catabolic process
positive regulation of mRNA 3'-end processing
positive regulation of histone phosphorylation
tissue regeneration
response to drug
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of mitotic cell cycle
response to DDT
positive regulation of fibroblast proliferation
digestive tract development
cell division
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
regulation of cell cycle
positive regulation of attachment of spindle microtubules to kinetochore
ventricular cardiac muscle cell development
positive regulation of cardiac muscle cell proliferation
regulation of chromosome condensation
cellular response to iron(III) ion
cellular response to fatty acid
cellular response to organic cyclic compound
cellular response to hypoxia
regulation of mitotic cell cycle spindle assembly checkpoint
histone H3-S10 phosphorylation involved in chromosome condensation
cyclin-dependent protein serine/threonine kinase activity
patched binding
protein binding
protein kinase binding
histone kinase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05003892.hg.1 | TC05000298.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003892.hg.1 | ENST00000256442.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003892.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000503507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000503507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000503507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000503507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000508407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000508407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000508407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000508407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000508407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000508407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000513102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000513102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000513102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000513102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | ENST00000513102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000503507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000503507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000503507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000503507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000507798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000508407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000508407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000508407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000508407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000508407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000508407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000513102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000513102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000513102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000513102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | ENST00000513102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007665.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151871.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151871.hg.1 | ENST00000503507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151871.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151871.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151871.hg.1 | ENST00000508407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151871.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151871.hg.1 | ENST00000256442.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151871.hg.1 | ENST00000503507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151871.hg.1 | ENST00000505500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151871.hg.1 | ENST00000506572.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151871.hg.1 | ENST00000508407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500151871.hg.1 | NM_031966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500151871.hg.1 | TC0500007665.hg.1 | Clariom D TC |

CCNB1
Genomic Map