Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023693.hg.1 |
ENST00000228280.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023693.hg.1 |
ENST00000347404.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023693.hg.1 |
ENST00000357116.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023693.hg.1 |
ENST00000378535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023693.hg.1 |
ENST00000537835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023693.hg.1 |
NM_000899.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023693.hg.1 |
NM_003994.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023693.hg.1 |
TC12001792.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200188770.hg.1 |
ENST00000228280.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188770.hg.1 |
ENST00000347404.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188770.hg.1 |
ENST00000357116.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188770.hg.1 |
ENST00000378535.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188770.hg.1 |
NM_000899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188770.hg.1 |
NM_003994.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188770.hg.1 |
ENST00000228280.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188770.hg.1 |
ENST00000347404.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188770.hg.1 |
ENST00000357116.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188770.hg.1 |
ENST00000378535.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188770.hg.1 |
NM_000899.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188770.hg.1 |
NM_003994.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188770.hg.1 |
TC1200011460.hg.1 |
Clariom D TC |