Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011305.hg.1 |
ENST00000408886.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011305.hg.1 |
ENST00000408886.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011305.hg.1 |
ENST00000623094.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011305.hg.1 |
ENST00000623094.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011305.hg.1 |
NR_122031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011305.hg.1 |
NR_122031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011305.hg.1 |
NR_122031.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011305.hg.1 |
NR_122031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188206.hg.1 |
ENST00000623094.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188206.hg.1 |
NR_122031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188206.hg.1 |
ENST00000623094.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188206.hg.1 |
NR_122031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188206.hg.1 |
TC1600011305.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600188205.hg.1 |
ENST00000408886.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188205.hg.1 |
ENST00000623094.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188205.hg.1 |
NR_122031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188205.hg.1 |
ENST00000408886.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188205.hg.1 |
ENST00000623094.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188205.hg.1 |
NR_122031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188205.hg.1 |
TC1600011305.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600188207.hg.1 |
NR_122031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188207.hg.1 |
NR_122031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188207.hg.1 |
TC1600011305.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011305.hg.1 |
ENST00000408886.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011305.hg.1 |
ENST00000408886.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011305.hg.1 |
ENST00000623094.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011305.hg.1 |
ENST00000623094.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011305.hg.1 |
NR_122031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011305.hg.1 |
NR_122031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011305.hg.1 |
NR_122031.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011305.hg.1 |
NR_122031.1 |
Refseq |