Assay Details
Gene Symbol: SNX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013321.3 | NP_037453.1 | 8 - 9 | 1042 | 73 | |
XM_011515329.2 | XP_011513631.1 | 8 - 9 | 1155 | 73 | ||
XM_011515330.1 | XP_011513632.1 | 8 - 9 | 1044 | 73 | ||
XM_017012083.1 | XP_016867572.1 | 8 - 9 | 1112 | 73 | ||
XM_017012084.1 | XP_016867573.1 | 8 - 9 | 1154 | 73 | ||
GenBank mRNA | AF121858.1 | - | 8 - 9 | 998 | 73 | |
AK074544.1 | - | 8 - 9 | 991 | 73 | ||
BC007785.2 | - | 8 - 9 | 971 | 73 | ||
BC021565.1 | - | 8 - 9 | 1031 | 4564502 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001096.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07016890.hg.1 | TC07001096.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07016890.hg.1 | ENST00000222990.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07016890.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000419693.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000419693.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000617724.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NR_106895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000419693.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000419693.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000617724.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NR_106895.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199737.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199737.hg.1 | ENST00000480807.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199737.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199737.hg.1 | ENST00000222990.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199737.hg.1 | ENST00000480807.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199737.hg.1 | NM_013321.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199737.hg.1 | TC0700013502.hg.1 | Clariom D TC |
