Assay Details
| Gene Symbol: SHROOM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020717.3 | NP_065768.2 | 6 - 7 | 3790 | 61 | |
| NR_027121.1 | 6 - 7 | 3790 | 61 | |||
| XM_017029682.1 | XP_016885171.1 | 7 - 8 | 4010 | 61 | ||
| XM_017029683.1 | XP_016885172.1 | 6 - 7 | 3875 | 61 | ||
| XM_017029684.1 | XP_016885173.1 | 6 - 7 | 4027 | 61 | ||
| XM_017029685.1 | XP_016885174.1 | 7 - 8 | 4027 | 61 | ||
| XM_017029686.1 | XP_016885175.1 | 5 - 6 | 3892 | 61 | ||
| GenBank mRNA | AB033028.1 | - | 6 - 7 | 3805 | 61 | |
| BC151240.1 | - | 6 - 7 | 3816 | 8860513 | 61 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001039.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013992.hg.1 | TC0X001039.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013992.hg.1 | ENST00000289292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013992.hg.1 | ENST00000376020.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013992.hg.1 | ENST00000460112.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013992.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013992.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013991.hg.1 | TC0X001039.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013991.hg.1 | ENST00000289292.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013991.hg.1 | ENST00000376020.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013991.hg.1 | ENST00000460112.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013991.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013991.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000483955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000483955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000483955.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000484922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | ENST00000484922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000483955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000483955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000483955.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000484922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | ENST00000484922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009684.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166190.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166190.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166190.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166190.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166190.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166190.hg.1 | ENST00000289292.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166190.hg.1 | ENST00000376020.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166190.hg.1 | ENST00000460112.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00166190.hg.1 | NM_020717.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166190.hg.1 | NR_027121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00166190.hg.1 | TC0X00009684.hg.1 | Clariom D TC |

SHROOM4
Genomic Map