Assay Details
| Gene Symbol: SWT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105518.1 | NP_001098988.1 | 14 - 15 | 2229 | 158 | |
| NM_017673.6 | NP_060143.4 | 14 - 15 | 2280 | 158 | ||
| XM_005245271.1 | XP_005245328.1 | 14 - 15 | 2287 | 158 | ||
| XM_005245273.1 | XP_005245330.1 | 14 - 15 | 2188 | 158 | ||
| XM_011509657.1 | XP_011507959.1 | 14 - 15 | 2267 | 158 | ||
| XM_017001527.1 | XP_016857016.1 | 14 - 15 | 2167 | 158 | ||
| XM_017001528.1 | XP_016857017.1 | 14 - 15 | 2287 | 158 | ||
| XM_017001529.1 | XP_016857018.1 | 15 - 16 | 2492 | 158 | ||
| XM_017001530.1 | XP_016857019.1 | 14 - 15 | 2287 | 158 | ||
| GenBank mRNA | AF288392.1 | - | 13 - 14 | 2126 | 158 | |
| AK000128.1 | - | 2 - 3 | 279 | 158 | ||
| AK093263.1 | - | 8 - 9 | 992 | 158 | ||
| AK225238.1 | - | 2 - 3 | 279 | 158 | ||
| AK291443.1 | - | 14 - 15 | 2146 | 158 | ||
| AK315802.1 | - | 14 - 15 | 2247 | 158 | ||
| BC030781.2 | - | 14 - 15 | 2282 | 4821611 | 158 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001596.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01024750.hg.1 | TC01001596.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024750.hg.1 | ENST00000367500.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024750.hg.1 | ENST00000367501.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024750.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024750.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180192.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180192.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180192.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180192.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180192.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180192.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180192.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180192.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180192.hg.1 | TC0100010905.hg.1 | Clariom D TC |

SWT1
Genomic Map