Assay Details
| Gene Symbol: SWT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105518.1 | NP_001098988.1 | 5 - 6 | 1079 | 109 | |
| NM_017673.6 | NP_060143.4 | 5 - 6 | 1130 | 109 | ||
| XM_005245271.1 | XP_005245328.1 | 5 - 6 | 1116 | 109 | ||
| XM_005245273.1 | XP_005245330.1 | 5 - 6 | 1017 | 109 | ||
| XM_011509657.1 | XP_011507959.1 | 5 - 6 | 1096 | 109 | ||
| XM_017001527.1 | XP_016857016.1 | 5 - 6 | 1017 | 109 | ||
| XM_017001528.1 | XP_016857017.1 | 5 - 6 | 1116 | 109 | ||
| XM_017001529.1 | XP_016857018.1 | 6 - 7 | 1321 | 109 | ||
| XM_017001530.1 | XP_016857019.1 | 5 - 6 | 1116 | 109 | ||
| GenBank mRNA | AF288392.1 | - | 4 - 5 | 976 | 109 | |
| AK127980.1 | - | 5 - 6 | 1017 | 109 | ||
| AK291443.1 | - | 5 - 6 | 996 | 109 | ||
| AK315802.1 | - | 5 - 6 | 1097 | 109 | ||
| BC030781.2 | - | 5 - 6 | 1132 | 4821611 | 109 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001596.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01024741.hg.1 | TC01001596.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024741.hg.1 | ENST00000367500.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024741.hg.1 | ENST00000367501.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024741.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024741.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180173.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180173.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180173.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180173.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180173.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180173.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180173.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180173.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180173.hg.1 | TC0100010905.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367500.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000367501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | ENST00000450350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_001105518.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010905.hg.1 | NM_017673.6 | Refseq |

SWT1
Genomic Map