Assay Details
Gene Symbol: IZUMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031735.2 | NP_001026905.2 | 7 - 8 | 613 | 87 | |
GenBank mRNA | AY753639.1 | - | 7 - 8 | 572 | 87 | |
BC014609.2 | - | 7 - 8 | 567 | 4825119 | 87 | |
DQ196309.1 | - | 7 - 8 | 600 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
