Assay Details
Gene Symbol: PREX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020820.3 | NP_065871.2 | 6 - 7 | 810 | 59 | |
GenBank mRNA | AB037836.2 | - | 6 - 7 | 673 | 59 | |
AJ320261.1 | - | 6 - 7 | 810 | 59 | ||
BC053616.1 | - | 2 - 3 | 260 | 59 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000920.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
superoxide metabolic process
G-protein coupled receptor signaling pathway
actin filament polymerization
T cell differentiation
neutrophil chemotaxis
regulation of actin filament polymerization
regulation of Rho protein signal transduction
intracellular signal transduction
neutrophil activation
positive regulation of GTPase activity
regulation of dendrite development
positive regulation of substrate adhesion-dependent cell spreading
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding
phospholipid binding
enzyme binding
superoxide metabolic process
G-protein coupled receptor signaling pathway
actin filament polymerization
T cell differentiation
neutrophil chemotaxis
regulation of actin filament polymerization
regulation of Rho protein signal transduction
intracellular signal transduction
neutrophil activation
positive regulation of GTPase activity
regulation of dendrite development
positive regulation of substrate adhesion-dependent cell spreading
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding
phospholipid binding
enzyme binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000371941.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000482556.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000496915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000496915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000496915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000496915.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | ENST00000620554.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009357.hg.1 | NM_020820.3 | Refseq |
