Assay Details
| Gene Symbol: RETSAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC011418.1 | - | 6 - 7 | 886 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200008243.hg.1 | ENST00000501107.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008243.hg.1 | ENST00000501107.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008243.hg.1 | ENST00000501107.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008243.hg.1 | ENST00000501107.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008243.hg.1 | ENST00000501107.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008243.hg.1 | ENST00000501107.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008243.hg.1 | ENST00000501107.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000490291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000490291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | ENST00000490291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000295802.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000409984.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000429806.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000438611.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000449375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000482694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000490291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000490291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | ENST00000490291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013261.hg.1 | NM_017750.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008243.hg.1 | ENST00000501107.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008243.hg.1 | ENST00000501107.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008243.hg.1 | ENST00000501107.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008243.hg.1 | ENST00000501107.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008243.hg.1 | ENST00000501107.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008243.hg.1 | ENST00000501107.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008243.hg.1 | ENST00000501107.2 | Ensembl |

RETSAT
Genomic Map