Assay Details
| Gene Symbol: WRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK297186.1 | - | 2 - 3 | 938 | 95 | |
| AK310268.1 | - | 2 - 3 | 869 | 95 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380713.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380713.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380713.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380713.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000380713.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000442773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000442773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000442773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000471468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000471468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000471468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000471468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000471468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000487869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000487869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000487869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000487869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000487869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000487869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000490860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000490860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000490860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | ENST00000490860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000333781.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380708.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380713.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380713.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380713.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380713.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000380713.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000398753.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000442773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000442773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000442773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000466787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000471468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000471468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000471468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000471468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000471468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000487869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000487869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000487869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000487869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000487869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000487869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000490860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000490860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000490860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | ENST00000490860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_001146218.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_004627.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008517.hg.1 | NM_004627.5 | Refseq |

WRB
Genomic Map