Assay Details
| Gene Symbol: HK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033500.2 | NP_277035.2 | 3 - 4 | 386 | 120 | |
| XM_011539732.1 | XP_011538034.1 | 3 - 4 | 293 | 120 | ||
| GenBank mRNA | AF073786.2 | - | 2 - 3 | 251 | 120 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF073786.2, NM_033500.2, XM_011539732.1 transcripts.
Set Membership: |
|

HK1
Genomic Map