Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005269.hg.1 |
TC07000387.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005269.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005269.hg.1 |
ENST00000545510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005269.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005269.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005271.hg.1 |
TC07000387.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005271.hg.1 |
ENST00000319636.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005271.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005271.hg.1 |
ENST00000527278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005271.hg.1 |
ENST00000545510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005271.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005271.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007579.hg.1 |
ENST00000600766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007579.hg.1 |
ENST00000600766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007579.hg.1 |
ENST00000600766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007579.hg.1 |
ENST00000600766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007579.hg.1 |
ENST00000600766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007579.hg.1 |
ENST00000600766.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005267.hg.1 |
TC07000387.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005267.hg.1 |
ENST00000319636.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005267.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005267.hg.1 |
ENST00000545510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005267.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005267.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005270.hg.1 |
TC07000387.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005270.hg.1 |
ENST00000319636.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005270.hg.1 |
ENST00000527278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005270.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005275.hg.1 |
TC07000387.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005275.hg.1 |
ENST00000319636.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005275.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005275.hg.1 |
ENST00000527278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005275.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005275.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005268.hg.1 |
TC07000387.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005268.hg.1 |
ENST00000319636.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005268.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005268.hg.1 |
ENST00000545510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005268.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005268.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005272.hg.1 |
TC07000387.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005272.hg.1 |
ENST00000319636.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005272.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005272.hg.1 |
ENST00000527278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005272.hg.1 |
ENST00000545510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005272.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005272.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197003.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197003.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197003.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197003.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197003.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197003.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197003.hg.1 |
TC0700013375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700197007.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197007.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197007.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197007.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197007.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197007.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197007.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197007.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197007.hg.1 |
TC0700013375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013375.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196999.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196999.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196999.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196999.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196999.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196999.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196999.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196999.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196999.hg.1 |
TC0700013375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700197002.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197002.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197002.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197002.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197002.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197002.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197002.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197002.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197002.hg.1 |
TC0700013375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1900007579.hg.1 |
ENST00000600766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007579.hg.1 |
ENST00000600766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007579.hg.1 |
ENST00000600766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007579.hg.1 |
ENST00000600766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007579.hg.1 |
ENST00000600766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007579.hg.1 |
ENST00000600766.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197001.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197001.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197001.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197001.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197001.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197001.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197001.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197001.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197001.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197001.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197001.hg.1 |
TC0700013375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700197000.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197000.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197000.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197000.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197000.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197000.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197000.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197000.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197000.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197000.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197000.hg.1 |
TC0700013375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700197004.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197004.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197004.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197004.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197004.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197004.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197004.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197004.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197004.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197004.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197004.hg.1 |
TC0700013375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700197005.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197005.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197005.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197005.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197005.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197005.hg.1 |
ENST00000395375.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197005.hg.1 |
ENST00000476120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197005.hg.1 |
ENST00000489672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700197005.hg.1 |
NM_021148.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197005.hg.1 |
NR_003099.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700197005.hg.1 |
TC0700013375.hg.1 |
Clariom D TC |