Assay Details
| Gene Symbol: C6orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025260.3 | NP_079536.2 | 4 - 5 | 582 | 154 | |
| NM_138277.2 | NP_612121.1 | 4 - 5 | 582 | 154 | ||
| XM_011514921.2 | XP_011513223.1 | 4 - 5 | 616 | 154 | ||
| GenBank mRNA | AJ292259.1 | - | 4 - 5 | 542 | 154 | |
| AJ292265.1 | - | 4 - 5 | 542 | 154 | ||
| AK092392.1 | - | 4 - 5 | 553 | 154 | ||
| BC113719.1 | - | 4 - 5 | 582 | 8322771 | 154 | |
| BC113721.1 | - | 4 - 5 | 582 | 8322773 | 154 | |
More Information
Set Membership: |
|

C6orf25
Genomic Map