Assay Details
Gene Symbol: MKI67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145966.1 | NP_001139438.1 | 11 - 12 | 1531 | 148 | |
NM_002417.4 | NP_002408.3 | 12 - 13 | 2611 | 148 | ||
XM_011539818.2 | XP_011538120.1 | 9 - 10 | 2342 | 148 | ||
GenBank mRNA | AJ567756.1 | - | 8 - 9 | 1141 | 148 | |
X65550.1 | - | 12 - 13 | 2612 | 148 | ||
X65551.1 | - | 11 - 12 | 1532 | 148 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001756.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
