Assay Details
Gene Symbol: MKI67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145966.1 | NP_001139438.1 | 2 - 3 | 285 | 77 | |
NM_002417.4 | NP_002408.3 | 2 - 3 | 285 | 77 | ||
GenBank mRNA | X65550.1 | - | 2 - 3 | 286 | 77 | |
X65551.1 | - | 2 - 3 | 286 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001756.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
DNA metabolic process
meiotic nuclear division
cell proliferation
response to organic cyclic compound
hyaluronan metabolic process
organ regeneration
cellular response to heat
hepatocyte proliferation
cholangiocyte proliferation
protein binding
ATP binding
protein C-terminus binding
poly(A) RNA binding
DNA metabolic process
meiotic nuclear division
cell proliferation
response to organic cyclic compound
hyaluronan metabolic process
organ regeneration
cellular response to heat
hepatocyte proliferation
cholangiocyte proliferation
protein binding
ATP binding
protein C-terminus binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368653.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000368654.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000464771.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000464771.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000478293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000478293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000478293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000478293.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000484853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000484853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000484853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000484853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | ENST00000617118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_001145966.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012235.hg.1 | NM_002417.4 | Refseq |
