Assay Details
Gene Symbol: MKI67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002417.4 | NP_002408.3 | 7 - 8 | 1674 | 93 | |
XM_011539818.2 | XP_011538120.1 | 4 - 5 | 1405 | 93 | ||
GenBank mRNA | BU855841.1 | - | 1 - 2 | 117 | 93 | |
X65550.1 | - | 7 - 8 | 1675 | 93 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001756.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
