Assay Details
Gene Symbol: ST6GALNAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006456.2 | NP_006447.2 | 6 - 7 | 1029 | 73 | |
GenBank mRNA | AJ251053.1 | - | 6 - 7 | 813 | 73 | |
AK225943.1 | - | 6 - 7 | 827 | 73 | ||
BC040455.1 | - | 6 - 7 | 811 | 4252792 | 73 | |
BT019972.1 | - | 6 - 7 | 774 | 73 | ||
U14550.1 | - | 6 - 7 | 813 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001903.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17025820.hg.1 | ENST00000225276.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17025820.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17025820.hg.1 | TC17001903.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700196497.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196497.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196497.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196497.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196497.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700196497.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196497.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196497.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196497.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196497.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700196497.hg.1 | TC1700011808.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
