Assay Details
Gene Symbol: ST6GALNAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006456.2 | NP_006447.2 | 7 - 8 | 1108 | 100 | |
GenBank mRNA | AJ251053.1 | - | 7 - 8 | 892 | 100 | |
AK225943.1 | - | 7 - 8 | 906 | 100 | ||
BC040455.1 | - | 7 - 8 | 890 | 4252792 | 100 | |
BT019972.1 | - | 7 - 8 | 853 | 100 | ||
U14550.1 | - | 7 - 8 | 892 | 100 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001903.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17025819.hg.1 | ENST00000225276.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17025819.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17025819.hg.1 | TC17001903.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700196494.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196494.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196494.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196494.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196494.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700196494.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196494.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196494.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196494.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700196494.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700196494.hg.1 | TC1700011808.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000225276.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000585736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000586520.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588005.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588120.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000588920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000589997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592508.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | ENST00000592979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011808.hg.1 | NM_006456.2 | Refseq |
