Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005189.hg.1 |
ENST00000288304.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005189.hg.1 |
ENST00000309117.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005189.hg.1 |
ENST00000398998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005189.hg.1 |
ENST00000399000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005189.hg.1 |
ENST00000399001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005189.hg.1 |
ENST00000476950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005189.hg.1 |
ENST00000539844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005189.hg.1 |
NM_006052.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005189.hg.1 |
TC21000441.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
ENST00000309117.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
ENST00000398998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
ENST00000399000.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
ENST00000399001.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
ENST00000476950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
ENST00000488368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
ENST00000495858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
NM_006052.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
TC2100008098.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
ENST00000309117.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
ENST00000398998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
ENST00000399000.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
ENST00000399001.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
ENST00000476950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
ENST00000488368.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
ENST00000495858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154686.hg.1 |
NM_006052.1 |
Refseq |