Assay Details
| Gene Symbol: MRPS18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193343.1 | NP_001180272.1 | 4 - 5 | 406 | 112 | |
| GenBank mRNA | AK300757.1 | - | 4 - 5 | 377 | 112 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001755.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06021109.hg.1 | TC06001755.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021109.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021109.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021111.hg.1 | TC06001755.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021111.hg.1 | ENST00000372133.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021111.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021111.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021112.hg.1 | TC06001755.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021112.hg.1 | ENST00000372118.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021112.hg.1 | ENST00000372133.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021112.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021112.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021114.hg.1 | TC06001755.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021114.hg.1 | ENST00000372116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021114.hg.1 | ENST00000372118.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021114.hg.1 | ENST00000372133.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021114.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021114.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000372116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000372116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000372116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000372116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000372116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000372116.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000427312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000427312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000427312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000427312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000427312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000427312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | ENST00000427312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021113.hg.1 | TC06001755.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021113.hg.1 | ENST00000372116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021113.hg.1 | ENST00000372118.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021113.hg.1 | ENST00000372133.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021113.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021113.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181741.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181741.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181741.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181741.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181741.hg.1 | TC0600011884.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000372116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000372116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000372116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000372116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000372116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000372116.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000372133.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000427312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000427312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000427312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000427312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000427312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000427312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | ENST00000427312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_001193343.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_018135.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011884.hg.1 | NM_018135.3 | Refseq |

MRPS18A
Genomic Map