Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015845.hg.1 |
ENST00000345330.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015845.hg.1 |
ENST00000351217.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015845.hg.1 |
ENST00000542234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015845.hg.1 |
ENST00000545878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015845.hg.1 |
NM_001161363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015845.hg.1 |
NM_001161364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015845.hg.1 |
NM_012428.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015845.hg.1 |
NM_017455.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15015845.hg.1 |
TC15001634.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
ENST00000345330.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
ENST00000351217.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
ENST00000561898.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
ENST00000562924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
ENST00000563691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
ENST00000565325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
NM_001161363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
NM_001161364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
NM_012428.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
NM_017455.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
ENST00000345330.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
ENST00000351217.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
ENST00000561898.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
ENST00000562924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
ENST00000563691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
ENST00000565325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
NM_001161363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
NM_001161364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
NM_012428.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
NM_017455.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500174278.hg.1 |
TC1500009984.hg.1 |
Clariom D TC |