Assay Details
| Gene Symbol: QSOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181701.3 | NP_859052.3 | 8 - 9 | 1120 | 93 | |
| GenBank mRNA | AK128077.1 | - | 8 - 9 | 785 | 93 | |
| AK128750.1 | - | 2 - 3 | 268 | 93 | ||
| AL834369.1 | - | 5 - 6 | 611 | 93 | ||
| BC047604.2 | - | 9 - 10 | 1002 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001726.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK128750.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09022053.hg.1 | TC09001726.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022053.hg.1 | ENST00000358701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022053.hg.1 | ENST00000389471.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022053.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022054.hg.1 | TC09001726.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022054.hg.1 | ENST00000358701.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022054.hg.1 | ENST00000389471.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09022054.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181866.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181866.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181866.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181866.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181866.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181866.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181866.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181866.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181866.hg.1 | TC0900011970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011970.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181867.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181867.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181867.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181867.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181867.hg.1 | ENST00000358701.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181867.hg.1 | ENST00000455222.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181867.hg.1 | ENST00000616829.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181867.hg.1 | NM_181701.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181867.hg.1 | TC0900011970.hg.1 | Clariom D TC |

QSOX2
Genomic Map