Assay Details
| Gene Symbol: SEC24A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252231.1 | NP_001239160.1 | 9 - 10 | 1790 | 129 | |
| NM_021982.2 | NP_068817.1 | 9 - 10 | 1790 | 129 | ||
| XM_006714523.2 | XP_006714586.1 | 8 - 9 | 1991 | 129 | ||
| XM_017008961.1 | XP_016864450.1 | 9 - 10 | 1435 | 129 | ||
| XM_017008962.1 | XP_016864451.1 | 9 - 10 | 1207 | 129 | ||
| XM_017008963.1 | XP_016864452.1 | 8 - 9 | 1991 | 129 | ||
| GenBank mRNA | AJ131244.1 | - | 9 - 10 | 1453 | 129 | |
| AK304060.1 | - | 9 - 10 | 1779 | 129 | ||
| BC019341.1 | - | 9 - 10 | 1751 | 3355949 | 129 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000665.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017008962.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
antigen processing and presentation of peptide antigen via MHC class I
intracellular protein transport
ER to Golgi vesicle-mediated transport
antigen processing and presentation of exogenous peptide antigen via MHC class II
regulation of low-density lipoprotein particle receptor biosynthetic process
COPII vesicle coating
positive regulation of protein secretion
positive regulation of cholesterol homeostasis
molecular_function
protein binding
zinc ion binding
antigen processing and presentation of peptide antigen via MHC class I
intracellular protein transport
ER to Golgi vesicle-mediated transport
antigen processing and presentation of exogenous peptide antigen via MHC class II
regulation of low-density lipoprotein particle receptor biosynthetic process
COPII vesicle coating
positive regulation of protein secretion
positive regulation of cholesterol homeostasis
molecular_function
protein binding
zinc ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05009202.hg.1 | TC05000665.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05009202.hg.1 | ENST00000322887.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05009202.hg.1 | ENST00000398844.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05009202.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05009202.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500159734.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500159734.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500159734.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500159734.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500159734.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500159734.hg.1 | ENST00000322887.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500159734.hg.1 | ENST00000398844.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500159734.hg.1 | ENST00000513123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500159734.hg.1 | NM_001252231.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500159734.hg.1 | NM_021982.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500159734.hg.1 | TC0500008700.hg.1 | Clariom D TC |

SEC24A
Genomic Map