Assay Details
| Gene Symbol: ADCY9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK022108.1 | - | 1 | 2013 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102724927 | LOC102724927 | -31.3kb | Sense | Chr. 16 - 3947609 - 3950444 on NCBI Build GRCh38 | |
| Downstream | sarcalumenin | SRL | 205.2kb | Sense | Chr. 16 - 4189374 - 4242253 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000294016.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000571889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000572288.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000574721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000575550.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | ENST00000576936.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009210.hg.1 | NM_001116.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 409573 | - | - |

ADCY9
Genomic Map