Assay Details
Gene Symbol: FAM101B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182705.2 | NP_874364.1 | 2 | 3215 | 105 | |
GenBank mRNA | AK093557.1 | - | 1 | 105 | ||
BC014203.2 | - | 1 | 271 | 105 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734177 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734165 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186326 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734188 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734186 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186320 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186327 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734200 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186290 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734159 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186301 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186316 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734157 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734163 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734162 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734156 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186324 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734176 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186288 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734173 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186300 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734193 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734189 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186318 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186323 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186322 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16932844 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186331 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734180 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186308 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186304 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186312 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734169 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734150 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734197 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186307 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734181 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30734154 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10186291 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 73190 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 705722 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 702304 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 354388 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P299318 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P299318 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P32151 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 8091 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 23361 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P104344 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P104344 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 35194 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P299318 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P299318 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10186323 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10186327 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10186324 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10186328 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10186331 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30734188 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30734193 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10186329 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30734200 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30734197 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10186322 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10186323 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30734188 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10186322 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10186328 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30734193 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10186327 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10186324 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30734200 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10186329 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30734197 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10186331 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P32151 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P299318 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P104344 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10186331 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10186330 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10186329 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10186327 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10186333 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10186321 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10186325 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10186322 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16932846 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16932847 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10186301 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10186314 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10186315 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10186318 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16932849 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16932845 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10186317 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16932850 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10186299 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10186300 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10186316 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P10186319 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P104344 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P104344 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P32151 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P299318 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 138784 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 47748 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20561896 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03200054 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186327 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186305 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186295 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186319 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186314 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186288 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186329 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186304 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186333 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186291 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186289 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186294 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186321 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186290 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186312 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186322 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186307 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186316 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186297 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16932847 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186325 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186310 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186299 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16932848 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10186331 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03200054 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_182705_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8010916 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8010917 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8010915 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 226876_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 226905_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11724993_at | - | - |
Affymetrix Human Genome U219 Array | 11724994_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11724993_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11724994_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1700009318.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009318.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009318.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009318.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009318.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009318.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009318.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009318.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009318.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009318.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009318.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009318.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700171061.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700171061.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700171061.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700171061.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700171061.hg.1 | TC1700009318.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700171060.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700171060.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700171060.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700171060.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700171060.hg.1 | TC1700009318.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700009318.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009318.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009318.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009318.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009318.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009318.hg.1 | ENST00000329099.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009318.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009318.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009318.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009318.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009318.hg.1 | NM_182705.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009318.hg.1 | NM_182705.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 226876_at | - | - |
Human Genome U133 Plus 2.0 Array | 226876_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 226905_at | - | - |
Human Genome U133 Plus 2.0 Array | 226905_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226905_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226905_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226905_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226905_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226876_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226876_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226905_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226905_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226876_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226905_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226876_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226876_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226876_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226876_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226905_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226876_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226876_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226876_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226905_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226905_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226905_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 226876_at2 | - | - |
Human Genome U95 Set | 59095_PM_at | - | - |
Human Genome U95 Set | 59095_at | - | - |
Human Genome U95 Set | 57616_PM_at | - | - |
Human Genome U95 Set | 57616_at | - | - |
Human Genome U133 Set | 226876_at | - | - |
Human Genome U133 Set | 226876_PM_at | - | - |
Human Genome U133 Set | 226905_PM_at | - | - |
Human Genome U133 Set | 226905_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054963 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A054963 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 103928 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1714065 | - | - |
HumanHT-12 v4 Expression BeadChip | ILMN_1714418 | BC031341.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1714418 | NM_182705.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1714418 | BC031341.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1714418 | NM_182705.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1714065 | - | - |
HumanHT-12 V4.0 expression beadchip | ILMN_1714418 | BC031341.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1714418 | NM_182705.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1714418 | BC031341.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1714418 | NM_182705.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1714418 | BC031341.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1714418 | NM_182705.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_33186900-S | BC031341.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_33186900-S | NM_182705.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_33186900-S | AF085987.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_1388-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0005050463 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0005050463 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1714418 | BC031341.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1714418 | NM_182705.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1714065 | - | - |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1714418 | BC031341.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1714418 | NM_182705.2 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 940246 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_1388 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1714418 | BC031341.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1714418 | NM_182705.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1714418 | BC031341.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1714418 | NM_182705.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1714065 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1714065 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1714418 | BC031341.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1714418 | NM_182705.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1714418 | BC031341.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1714418 | NM_182705.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1714418 | BC031341.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1714418 | NM_182705.2 | human_transcriptome |
