Assay Details
Gene Symbol: KIF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006612.5 | NP_006603.2 | 15 - 16 | 1774 | 94 | |
XM_005256424.2 | XP_005256481.1 | 16 - 17 | 1765 | 94 | ||
GenBank mRNA | AB014606.2 | - | 15 - 16 | 1770 | 94 | |
AK307401.1 | - | 15 - 16 | 1617 | 94 | ||
BC034993.1 | - | 15 - 16 | 1748 | 5266798 | 94 | |
U91329.1 | - | 14 - 15 | 1530 | 94 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000060.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000572959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000572959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17000708.hg.1 | ENST00000320785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17000708.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17000708.hg.1 | TC17000060.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000572959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000572959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700145775.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145775.hg.1 | ENST00000572959.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145775.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700145775.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145775.hg.1 | ENST00000572959.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145775.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700145775.hg.1 | TC1700006660.hg.1 | Clariom D TC |
