Assay Details
Gene Symbol: KIF1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006612.5 | NP_006603.2 | 1 - 2 | 214 | 140 | |
GenBank mRNA | AB014606.2 | - | 1 - 2 | 210 | 140 | |
BC034993.1 | - | 1 - 2 | 188 | 5266798 | 140 | |
BM458656.1 | - | 1 - 2 | 214 | 140 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000060.hg
Additional Information:
This assay is in the 5'UTR region of the AB014606.2,BC034993.1,NM_006612.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17000693.hg.1 | ENST00000320785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17000693.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17000693.hg.1 | TC17000060.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17000694.hg.1 | ENST00000320785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17000694.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17000694.hg.1 | TC17000060.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17000692.hg.1 | ENST00000320785.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17000692.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17000692.hg.1 | TC17000060.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000572959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000572959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700145756.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145756.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145756.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700145756.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145756.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145756.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700145756.hg.1 | TC1700006660.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700145758.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145758.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145758.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700145758.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145758.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145758.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700145758.hg.1 | TC1700006660.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000572959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000572959.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000573815.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006660.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700145755.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145755.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145755.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700145755.hg.1 | ENST00000320785.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145755.hg.1 | ENST00000574165.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700145755.hg.1 | NM_006612.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700145755.hg.1 | TC1700006660.hg.1 | Clariom D TC |
