Assay Details
Gene Symbol: ODF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014440.3 | NP_001014440.2 | 2 - 3 | 60 | ||
GenBank mRNA | AW291097.1 | - | 1 - 2 | 30 | 60 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22001501.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22015687.hg.1 | TC22001501.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22015687.hg.1 | ENST00000329363.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015687.hg.1 | ENST00000405135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015687.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015687.hg.1 | ENST00000438960.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015687.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22015693.hg.1 | TC22001501.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22015693.hg.1 | ENST00000329363.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015693.hg.1 | ENST00000405135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015693.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015693.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22015685.hg.1 | TC22001501.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22015685.hg.1 | ENST00000329363.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015685.hg.1 | ENST00000401779.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015685.hg.1 | ENST00000405135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015685.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015685.hg.1 | ENST00000438960.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22015685.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170820.hg.1 | TC2200009363.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200170820.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170820.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170820.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170820.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170820.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170820.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170820.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170820.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170820.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170820.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170817.hg.1 | TC2200009363.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200170817.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170817.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170817.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170817.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170817.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170817.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170817.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170817.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170817.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170817.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170817.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170817.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170819.hg.1 | TC2200009363.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200170819.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170819.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170819.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170819.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170819.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170819.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170819.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170819.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
