Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015672.hg.1 |
TC22001501.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015672.hg.1 |
ENST00000329363.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015672.hg.1 |
ENST00000401779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015672.hg.1 |
ENST00000403326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015672.hg.1 |
ENST00000405135.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015672.hg.1 |
ENST00000428989.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22015672.hg.1 |
NM_001014440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
TC2200009363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
ENST00000329363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
ENST00000401779.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
ENST00000403326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
ENST00000405135.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
ENST00000428989.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
ENST00000463472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
ENST00000469660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
NM_001014440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
ENST00000329363.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
ENST00000401779.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
ENST00000403326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
ENST00000405135.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
ENST00000428989.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
ENST00000463472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
ENST00000469660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200170811.hg.1 |
NM_001014440.3 |
Refseq |