Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009056.hg.1 |
ENST00000534436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003940.hg.1 |
TC05000300.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003940.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003940.hg.1 |
ENST00000512880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003940.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003935.hg.1 |
TC05000300.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003935.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003935.hg.1 |
ENST00000507022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003935.hg.1 |
ENST00000512880.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05003935.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010163.hg.1 |
ENST00000421291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000503793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000503793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000507022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000507022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000507022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151930.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151930.hg.1 |
ENST00000507022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151930.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151930.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151930.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151930.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151930.hg.1 |
ENST00000507022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151930.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151930.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151930.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151930.hg.1 |
TC0500007667.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500151935.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151935.hg.1 |
ENST00000503793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151935.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151935.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151935.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151935.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151935.hg.1 |
ENST00000503793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151935.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151935.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500151935.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500151935.hg.1 |
TC0500007667.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009056.hg.1 |
ENST00000534436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000256441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000503793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000503793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000507022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000507022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000507022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000512880.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
ENST00000602380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007667.hg.1 |
NM_033281.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010163.hg.1 |
ENST00000421291.1 |
Ensembl |