Assay Details
| Gene Symbol: TAF7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168474.1 | NP_001161946.1 | 9 - 10 | 806 | 100 | |
| NM_024885.3 | NP_079161.3 | 9 - 10 | 974 | 100 | ||
| GenBank mRNA | AF285595.1 | - | 9 - 10 | 808 | 100 | |
| AK308261.1 | - | 9 - 10 | 976 | 100 | ||
| BC043391.1 | - | 9 - 10 | 960 | 6154641 | 100 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001215.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
multicellular organism development
spermatogenesis
biological_process
cell differentiation
RNA polymerase II transcriptional preinitiation complex assembly
molecular_function
transcription coactivator activity
transcription factor binding
histone acetyltransferase binding
transcription regulatory region DNA binding
regulation of transcription from RNA polymerase II promoter
multicellular organism development
spermatogenesis
biological_process
cell differentiation
RNA polymerase II transcriptional preinitiation complex assembly
molecular_function
transcription coactivator activity
transcription factor binding
histone acetyltransferase binding
transcription regulatory region DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016352.hg.1 | TC0X001215.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016352.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016352.hg.1 | ENST00000372907.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016352.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X016352.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00170335.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00170335.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00170335.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00170335.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00170335.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00170335.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00170335.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00170335.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00170335.hg.1 | TC0X00010351.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000324762.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000356784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | ENST00000372907.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_001168474.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010351.hg.1 | NM_024885.3 | Refseq |

TAF7L
Genomic Map