Assay Details
| Gene Symbol: NLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005248559.2 | XP_005248616.1 | 9 - 10 | 1649 | 74 | |
| XM_017009673.1 | XP_016865162.1 | 10 - 11 | 1713 | 74 | ||
| GenBank mRNA | AK027602.1 | - | 3 - 4 | 664 | 74 | |
More Information
Set Membership: |
|

NLN
Genomic Map